Anti-Gelatin, Skin Rabbit polyclonal antibody
USBIG2024-05B
- Antibody type:Primary
- Antigen name:Gelatin, Skin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1:1000-1:10000
Type: Primary
Antigen: Gelatin, Skin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine