Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-FOS Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIF6040-20A-FITC
undefined
Anti-FOS Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIF6040-20A-FITC

 

  • Antibody type:
    Primary
  • Antigen name:
    FBJ murine osteosarcoma viral oncogene homolog
  • Antigen symbol:
    FOS
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Thr232
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Dot Blot, Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000 Dot Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human