Anti-Endothelial Cell Internalizing Surface Antigen Mouse monoclonal antibody [clone: GES-HB]
Catalog # USBIE3010-15
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Endothelial Cell Internalizing Surface Antigen
- Clonality:Monoclonal
- Clone:GES-HB
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:20
Type: Primary
Antigen: Endothelial Cell Internalizing Surface Antigen
Clonality: monoclonal
Clone: GES-HB
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human