Anti-DNA Polymerase, eta Mouse Polyclonal Antibody
Catalog # USBID3935-33K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:DNA Polymerase eta
- Antigen symbol:POLH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry/Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: DNA Polymerase, eta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human