Anti-DNA Polymerase, beta Rabbit Polyclonal Antibody
USBID3935-12D
- Antibody type:Primary
- Antigen name:DNA Polymerase beta
- Antigen symbol:POLB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:500
Type: Primary
Antigen: DNA Polymerase, beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse