Anti-C5b-9 Terminal Complement Complex Mouse monoclonal antibody [clone: 9B1]
USBIC7850-26
- Antibody type:Primary
- Antigen name:C5b-9 Terminal Complement Complex
- Clonality:Monoclonal
- Clone:9B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Pig,Horse,Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: C5b-9 Terminal Complement Complex
Clonality: Monoclonal
Clone: 9B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Horse, Human, Pig