Anti-COL2 Mouse Monoclonal Antibody [clone: 12F6]
USBIC7510-37A
- Antibody type:Primary
- Antigen name:collagen, type II
- Antigen symbol:COL2
- Clonality:Monoclonal
- Clone:12F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Bovine
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: Monoclonal
Clone: 12F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine