Anti-CEACAM 5,6 Mouse monoclonal antibody [clone: 5E505]
Catalog # USBIC2589-78
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CEACAM 5,6
- Clonality:Monoclonal
- Clone:5E505
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CEACAM 5,6
Clonality: Monoclonal
Clone: 5E505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human