Anti-BrdU Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B320]
USBIB2850-10H
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:10B320
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Flow Cytometry:1:10;
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone: 10B320
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: