Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-BrdU Sheep polyclonal antibody unconjugated
  USBIB2850-04B
Anti-BrdU Sheep polyclonal antibody unconjugated
  USBIB2850-04B

 

  • Antibody type:
    Primary
  • Antigen name:
    Bromodeoxyuridine
  • Antigen symbol:
    BrdU
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Cat
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBSbefore the addition of 40% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Immunoprecipitation: 25-100 ?g/ml

Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Cat