Anti-BrdU Mouse monoclonal antibody unconjugated [clone: 5F214]
USBIB2850-03A
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:5F214
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:10
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone: 5F214
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: