Anti-BrdU Mouse monoclonal antibody unconjugated [clone: 5F172]
Catalog # USBIB2850-03
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:5F172
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,4, 0,5% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:10
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone: 5F172
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: