Anti-BrdU Mouse Monoclonal Antibody (Biotin) [clone: 6D11]
Catalog # USBIB2850-01H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:6D11
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Form:liquid
- Storage buffer:300µl of PBS, pH 7,4, containing 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Size:60 Tests
- Pk:60 Tests
Specifications
About this item
Recommended dilution: Flow Cytometry:1:20
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone: 6D11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: