Anti-BRD2 Rabbit monoclonal antibody unconjugated [clone: D89B4]
Catalog # USBIB2735-01E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:bromodomain containing 2
- Antigen symbol:BRD2
- Clonality:Monoclonal
- Clone:D89B4
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: BRD2 (bromodomain containing 2)
Clonality: monoclonal
Clone: D89B4
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse