Anti-BRAM1 Rabbit polyclonal antibody unconjugated
USBIB2705-30A
- Antibody type:Primary
- Antigen symbol:BRAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: BRAM1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse