Anti-ANXA4 Rabbit Polyclonal Antibody
USBIA2295-15D5
- Antibody type:Primary
- Antigen name:Annexin A4
- Antigen symbol:ANXA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:Lipocortin IV
- Storage buffer:PBS, pH 7,4, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot:1-2?g/ml; Immunochemistry:1-2?g/ml
Type: Primary
Antigen: ANXA4 (annexin A4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rabbit, Rat