Anti-ACTN4 Mouse Monoclonal Antibody [clone: 4D10]
Catalog # USBIA0767-04F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:actinin alpha 4
- Antigen symbol:ACTN4
- Clonality:Monoclonal
- Clone:4D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:592 to 702
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 20 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: ACTN4 (actinin, alpha 4)
Clonality: monoclonal
Clone: 4D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human