Anti-GABA-B Receptor 2 Mouse Monoclonal Antibody [clone: S81-2]
USBI167587
- Antibody type:Primary
- Antigen name:GABA-B Receptor 2
- Clonality:Monoclonal
- Clone:S81-2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: GABA-B Receptor 2
Clonality: monoclonal
Clone: S81-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat