Anti-VSIG2 Rat Monoclonal Antibody [clone: 771625]
USBI145652
- Antibody type:Primary
- Antigen name:V-set and immunoglobulin domain containing 2
- Antigen symbol:VSIG2
- Clonality:Monoclonal
- Clone:771625
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Form:lyophilized powder
- Amino acid number:25 to 244
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 825 ?g/ml
Type: Primary
Antigen: VSIG2 (V-set and immunoglobulin domain containing 2)
Clonality: monoclonal
Clone: 771625
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse