Anti-Glutathione S-Transferase mu 1 Mouse Monoclonal Antibody [clone: 842516]
USBI145480
- Antibody type:Primary
- Antigen name:Glutathione S-Transferase mu 1
- Clonality:Monoclonal
- Clone:842516
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:1 to 218
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 µg/ml
Type: Primary
Antigen: Glutathione S-Transferase mu 1
Clonality: monoclonal
Clone: 842516
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human