Anti-Green Fluorescent Protein, enhanced EGFP, EYFP) Mouse Monoclonal Antibody [clone: 13k51]
Catalog # USBI143178
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Green Fluorescent Protein, enhanced EGFP, EYFP
- Antigen symbol:GFP
- Clonality:Monoclonal
- Clone:13k51
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Green Fluorescent Protein, enhanced EGFP, EYFP)
Clonality: monoclonal
Clone: 13k51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human