Anti-AURKA Mouse Monoclonal Antibody [clone: 13k13]
Catalog # USBI143140
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:aurora kinase A
- Antigen symbol:AURKA
- Clonality:Monoclonal
- Clone:13k13
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:ascites fluid.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:200
Type: Primary
Antigen: AURKA (aurora kinase A)
Clonality: monoclonal
Clone: 13k13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey