Anti-Lymphocyte Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI138099
- Antibody type:Primary
- Antigen name:Lymphocyte
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:2 mg
- Pk:2 mg
Type: Primary
Antigen: Lymphocyte
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat