Anti-ALYREF Rabbit Monoclonal Antibody [clone: 13G74]
Catalog # USBI137502
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Aly/REF export factor
- Antigen symbol:ALYREF
- Clonality:Monoclonal
- Clone:13G74
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol and <0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
- Recommended dilution: Immunofluorescence: 1:400; Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: ALYREF (Aly/REF export factor)
Clonality: monoclonal
Clone: 13G74
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat