Anti-ZBTB16 Mouse Monoclonal Antibody [clone: 3A7]
Catalog # USBI135507
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 16
- Antigen symbol:ZBTB16
- Clonality:Monoclonal
- Clone:3A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:QRELFSKLGELAVGMKSESRTIGEQCSVCGVELPDNEAVEQHRKLHSGMKTYGCELCGKRFLDSLRLRMHLLAHSAGAKAFVCDQCGAQFSKEDALETHR*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ZBTB16 (zinc finger and BTB domain containing 16)
Clonality: monoclonal
Clone: 3A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human