Anti-Tetratricopeptide Repeat Protein 8 Mouse Monoclonal Antibody [clone: 7E2]
Catalog # USBI134862
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Tetratricopeptide Repeat Protein 8
- Antigen symbol:TTC8
- Clonality:Monoclonal
- Clone:7E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:416 to 515
- Storage buffer:PBS, pH 7,2.
- Sequence:AHQCFRLALVNNNNHAEAYNNLAVLEMRKGHVEQARALLQTASSLAPHMYEPHFNFATISDKIGDLQRSYVAAQKSEAAFPDHVDTQHLIKQLRQHFAM
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Tetratricopeptide Repeat Protein 8
Clonality: monoclonal
Clone: 700
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat