Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-SPOCK3 Mouse Monoclonal Antibody [clone: 1A10]
  USBI133789
Anti-SPOCK3 Mouse Monoclonal Antibody [clone: 1A10]
  USBI133789

 

  • Antibody type:
    Primary
  • Antigen name:
    sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
  • Antigen symbol:
    SPOCK3
  • Clonality:
    Monoclonal
  • Clone:
    1A10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    liquid
  • Amino acid number:
    132 to 231
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    RGPILSTCKQCPVVYPSPVCGSDGHTYSFQCKLEYQACVLGKQISVKCEGHCPCPSDKPTSTSRNVKRACSDLEFREVANRLRDWFKALHESGSQNKKT
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: SPOCK3 (sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3)
Clonality: monoclonal
Clone: 1A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human