Order Entry
United Kingdom
ContactUsLinkComponent
Anti-AKAP17A Mouse Polyclonal Antibody
Anti-AKAP17A Mouse Polyclonal Antibody
  USBI133230
Anti-AKAP17A Mouse Polyclonal Antibody
  USBI133230

 

  • Antibody type:
    Primary
  • Antigen name:
    A kinase (PRKA) anchor protein 17
  • Antigen symbol:
    AKAP17A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MAAATIVHDTSEAVELCPAYGLYLKPITKMTISVALPQLKQPGKSISNWEVMERLKGMVQNHQFSTLRISKSTMDFIRFEGEVENKSLVKSFLACLDGKTIKLSGFSDILKVRAAEFKIDFPTRHDWDSFFRDAKDMNETLPGERPDTIHLEGLPCKWFALKESGSEKPSEDVLVKVFEKFGEIRNVDIPMLDSYREEMTGRNFHTFSFGGHLNFEAYVQYREYMGFIQAMSALRGMKLMYKGEDGKAVACNIKVSFDSTKHLSDASIKKRQLERQKLQELEQQREEQKRREKEAEERQRAEERKQKELEELERERKREEKLRKREQKQRDRELRRNQKKLEKLQAEEQKQLQEKIKLEERKLLLAQRNLQSIRLIAELLSRAKVPGGSLCSRQPRPGCPQCPPLKCGRRHGAVSPPAAAVATKPALMPRMTAPSREGVALVCRSR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: AKAP17A (A kinase (PRKA) anchor protein 17A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human