Anti-PIP5K1C Mouse Monoclonal Antibody [clone: 7D11]
Catalog # USBI131363
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylinositol-4-phosphate 5-kinase, type I, gamma
- Antigen symbol:PIP5K1C
- Clonality:Monoclonal
- Clone:7D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:561 to 668
- Storage buffer:PBS, pH 7,2.
- Sequence:RPQEEPPAEEDLQQITVQVEPACSVEIVVPKEEDAGVEASPAGASAAVEVETASQASDEEGAPASQASDEEDAPATDIYFPTDERSWVYSPLHYSAQAPPASDGESD*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PIP5K1C (phosphatidylinositol-4-phosphate 5-kinase, type I, gamma)
Clonality: monoclonal
Clone: 7D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human