Anti-HSD3B1 Mouse Monoclonal Antibody [clone: 3C11-D4]
USBI128104
- Antibody type:Primary
- Antigen name:hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
- Antigen symbol:HSD3B1
- Clonality:Monoclonal
- Clone:3C11-D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MTGWSCLVTGAGGFLGQRIIRLLVKEKELKEIRVLDKAFGPELREEFSKLQNKTKLTVLEGDILDEPFLKRACQDVSVIIHTACIIDVFGVTHRESIMNVNVKGTQLLLEACVQASVPVFIYTSSIEVAGPNSYKEIIQNGHEEEPLENTWPAPYPHSKKLAEKAVLAANGWNLKNGGTLYTCALRPMYIYGEGSRFLSASINEALNNNGILSSVGKFSTVNPVYVGNVAWAHILALRALQDPKKAPSIRGQFYYISDDTPHQSYDNLNYTLSKEFGLRLDSRWSFPLSLMYWIGFLLEIVSFLLRPIYTYRPPFNRHIVTLSNSVFTFSYKKAQRDLAYKPLYSWEEAKQKTVEWVGSLVDRHKENLKSKTQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone: 3C11-D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human