Anti-HSD3B1 Rabbit Polyclonal Antibody
Catalog # USBI128102
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
- Antigen symbol:HSD3B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Amino acid number:1 to 373
- Sequence:MTGWSCLVTGAGGFLGQRIIRLLVKEKELKEIRVLDKAFGPELREEFSKLQNKTKLTVLEGDILDEPFLKRACQDVSVIIHTACIIDVFGVTHRESIMNVNVKGTQLLLEACVQASVPVFIYTSSIEVAGPNSYKEIIQNGHEEEPLENTWPAPYPHSKKLAEKAVLAANGWNLKNGGTLYTCALRPMYIYGEGSRFLSASINEALNNNGILSSVGKFSTVNPVYVGNVAWAHILALRALQDPKKAPSIRGQFYYISDDTPHQSYDNLNYTLSKEFGLRLDSRWSFPLSLMYWIGFLLEIVSFLLRPIYTYRPPFNRHIVTLSNSVFTFSYKKAQRDLAYKPLYSWEEAKQKTVEWVGSLVDRHKETLKSKTQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human