Anti-HMGCS2 Mouse Monoclonal Antibody [clone: 1E9]
USBI127962
- Antibody type:Primary
- Antigen name:3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial)
- Antigen symbol:HMGCS2
- Clonality:Monoclonal
- Clone:1E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:424 to 508
- Storage buffer:PBS, pH 7,2.
- Sequence:RVSQDAAPGSPLDKLVSSTSDLPKRLASRKCVSPEEFTEIMNQREQFYHKVNFSPPGDTNSLFPGTWYLERVDEQHRRKYARRPV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HMGCS2 (3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial))
Clonality: monoclonal
Clone: 1000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human