Anti-CAMKV Mouse Polyclonal Antibody
Catalog # USBI124302
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CaM kinase-like vesicle-associated
- Antigen symbol:CAMKV
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 501
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CAMKV (CaM kinase-like vesicle-associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat