Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 1G6]
Catalog # USBI123196
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:aldehyde dehydrogenase 1 family, member A1
- Antigen symbol:ALDH1A1
- Clonality:Monoclonal
- Clone:1G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: monoclonal
Clone: 1G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human