Anti-DNAJB9 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI034706-AP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:DnaJ (Hsp40) homolog, subfamily B, member 9
- Antigen symbol:DNAJB9
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:39 to 69
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: DNAJB9 (DnaJ (Hsp40) homolog, subfamily B, member 9)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse