Anti-EXO5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI034579-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Exonuclease 5
- Antigen symbol:EXO5
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Hamster,Human,Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:173 to 201
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Type: Primary
Antigen: EXO5 (exonuclease 5)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse