Anti-AP4M1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI031952-PE
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 4, mu 1 subunit
- Antigen symbol:AP4M1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:222 to 251
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: AP4M1 (adaptor-related protein complex 4, mu 1 subunit)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human