Anti-EIF2AK3 Mouse Monoclonal Antibody [clone: 13B240]
Catalog # USBI030162
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 2-alpha kinase 3
- Antigen symbol:EIF2AK3
- Clonality:Monoclonal
- Clone:13B240
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: EIF2AK3 (eukaryotic translation initiation factor 2-alpha kinase 3)
Clonality: Monoclonal
Clone: 13B240
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human