Anti-HIST3H3 Rabbit Polyclonal Antibody
Catalog # USBI029499
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:histone cluster 3, H3
- Antigen symbol:HIST3H3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Modification:phospho-Thr11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:25 Tests
- Pk:25 Tests
Specifications
About this item
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human