Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-TLR10 Mouse Monoclonal Antibody [clone: 28M7A3]
  NOVUNBP2-27273
Anti-TLR10 Mouse Monoclonal Antibody [clone: 28M7A3]
  NOVUNBP2-27273

 

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like Receptor 10
  • Antigen symbol:
    TLR10
  • Clonality:
    Monoclonal
  • Clone:
    28M7A3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Gene ID:
    81793
  • Antigen synonyms:
    CD290
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: TLR10 (toll-like receptor 10)
Clonality: monoclonal
Clone: 28M7A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human