Anti-IL3RA Mouse Monoclonal Antibody [clone: 18N6B12]
Catalog # NOVUNBP2-27225
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 3 Receptor Alpha
- Antigen symbol:IL3RA
- Clonality:Monoclonal
- Clone:18N6B12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Gene ID:3563
- Antigen synonyms:IL3RX|IL3RY|CD123|hIL-3Ra|IL3RAY|IL3R
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: IL3RA (interleukin 3 receptor, alpha (low affinity))
Clonality: monoclonal
Clone: 18N6B12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human