Anti-Dectin 2 Mouse Monoclonal Antibody [clone: 3D1]
Catalog # NOVUNBP2-27159
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:C-type lectin domain family 6, member A
- Antigen symbol:CLEC6A
- Clonality:Monoclonal
- Clone:3D1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:93978
- Antigen synonyms:CLECSF10|CLEC4N
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Dectin 2 (dectin 2)
Clonality: monoclonal
Clone: 3D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human