Anti-S1PR2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-26691
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:sphingosine-1-phosphate receptor 2
- Antigen symbol:S1PR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Gene ID:9294
- Antigen synonyms:AGR16|H218|EDG-5|EDG5|LPB2|S1P2|Gpcr13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: S1PR2 (sphingosine-1-phosphate receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: