Anti-TLR6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 86B1153.2]
NOVUNBP2-24969
- Antibody type:Primary
- Antigen name:Toll-like Receptor 6
- Antigen symbol:TLR6
- Clonality:Monoclonal
- Clone:86B1153.2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:10333
- Antigen synonyms:CD286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: monoclonal
Clone: 86B1153.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human