Anti-NLRP10 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24606
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:NLR family, pyrin domain containing 10
- Antigen symbol:NLRP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:338322
- Antigen synonyms:NALP10|PYNOD|NOD8|CLR11.1|PAN5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NLRP10 (NLR family, pyrin domain containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human