Anti-SGK1 Rabbit Polyclonal Antibody
NOVUNBP2-19010
- Antibody type:Primary
- Antigen name:Serum/glucocorticoid regulated kinase 1
- Antigen symbol:SGK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:6446
- Antigen synonyms:SGK1|Serum/glucocorticoid-regulated kinase 1|SGK|Serine/threonine-protein kinase Sgk1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: SGK1 (serum/glucocorticoid regulated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human