Anti-MMP7 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Rabbit Polyclonal MMP7 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:300
Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Specifications
- Catalog No:
- NOVUNBP1-99123
- NOVUNBP1-99123SS
- Antigen symbol:
- MMP7
- MMP7
- Antigen name:
- Matrix Metalloproteinase 7
- Matrix Metalloproteinase 7
- Antigen synonyms:
- PUMP-1|MPSL1|MMP-7
- PUMP-1|MPSL1|MMP-7
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4316
- 4316
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together