Anti-PLK5 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNBP1-97306C
- Antibody type:Primary
- Antigen name:Polo-like kinase 5
- Antigen symbol:PLK5
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:126520
- Antigen synonyms:inactive serine/threonine-protein kinase PLK5|polo-like kinase 5 pseudogene|PLK-5|SgK384ps|PLK5P
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: PLK5 (polo-like kinase 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse