Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MFN1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 11E9-1H12]
Anti-MFN1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 11E9-1H12]
Catalog # NOVUNBP1-71775V
Supplier:  Novus Biologicals
Anti-MFN1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 11E9-1H12]
Catalog # NOVUNBP1-71775V
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    mitofusin 1
  • Antigen symbol:
    MFN1
  • Clonality:
    Monoclonal
  • Clone:
    11E9-1H12
  • Conjugation:
    DyLight® 405
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    55669
  • Antigen synonyms:
    hfzo1|hfzo2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: MFN1 (mitofusin 1)
Clonality: monoclonal
Clone: 11E9-1H12
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat