Anti-Oval Cell Marker Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: OC2-4E8]
Catalog # NOVUNBP1-18971F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-4E8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-4E8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse